Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000071.2 | NP_000062.1 | 4 | 531 | ||
NM_001178008.2 | NP_001171479.1 | 4 | 538 | |||
NM_001178009.2 | NP_001171480.1 | 4 | 538 | |||
NM_001320298.1 | NP_001307227.1 | 5 | 532 | |||
XM_011529777.1 | XP_011528079.1 | 4 | 474 | |||
XM_011529781.1 | XP_011528083.1 | 5 | 539 | |||
XM_011529782.1 | XP_011528084.1 | 4 | 937 | |||
XM_017028489.1 | XP_016883978.1 | 5 | 934 | |||
XM_017028490.1 | XP_016883979.1 | 5 | 931 | |||
XM_017028491.1 | XP_016883980.1 | 4 | 474 | |||
XM_017028492.1 | XP_016883981.1 | 5 | 539 | |||
GenBank mRNA | AK294608.1 | - | 6 | 403 | ||
AK301063.1 | - | 2 | 133 | |||
AK313691.1 | - | 4 | 436 | |||
AL137314.1 | - | 1 | 58 | |||
AL531222.3 | - | 4 | 522 | |||
BC000440.2 | - | 4 | 406 | 2820694 | ||
BC010242.1 | - | 4 | 455 | 4154798 | ||
BC011381.1 | - | 4 | 394 | 3869615 | ||
BT007154.1 | - | 2 | 286 | |||
DC300040.1 | - | 4 | 531 | |||
DC315340.1 | - | 5 | 532 | |||
L14577.1 | - | 4 | 444 | |||
L19501.1 | - | 4 | 467 | |||
X82166.1 | - | 4 | 433 |
Gene Symbol: CBSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321073.1 | NP_001308002.1 | 4 | 538 | ||
XM_011546094.1 | XP_011544396.1 | 4 | 430 | |||
XM_011546095.2 | XP_011544397.1 | 4 | 572 | |||
XM_011546097.2 | XP_011544399.1 | 5 | 599 | |||
XM_011546098.1 | XP_011544400.1 | 4 | 937 | |||
XM_011546099.1 | XP_011544401.1 | 4 | 430 | |||
XM_017028211.1 | XP_016883700.1 | 5 | 935 | |||
XM_017028212.1 | XP_016883701.1 | 6 | 991 | |||
XM_017028213.1 | XP_016883702.1 | 4 | 430 | |||
XM_017028214.1 | XP_016883703.1 | 5 | 932 | |||
XM_017028215.1 | XP_016883704.1 | 5 | 547 | |||
XM_017028216.1 | XP_016883705.1 | 4 | 937 | |||
XM_017028217.1 | XP_016883706.1 | 4 | 430 | |||
XM_017028218.1 | XP_016883707.1 | 5 | 599 | |||
GenBank mRNA | BC007257.1 | - | 4 | 508 | 3028099 |