Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCOF1 SIRNA Assays ›
Gene Symbol:
TCOF1
Gene Name:
treacle ribosome biogenesis factor 1
Gene Aliases:
MFD1, TCS, TCS1, treacle
Chromosome Location:
Chr.5: 150357639 - 150400308 on Build GRCh38
UniGene ID:
Hs.519672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529267
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCOF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000356.3
NP_000347.2
14
2280
NM_001008657.2
NP_001008657.1
15
2511
NM_001135243.1
NP_001128715.1
15
2511
NM_001135244.1
NP_001128716.1
15
2511
NM_001135245.1
NP_001128717.1
14
2280
NM_001195141.1
NP_001182070.1
15
2511
XM_005268502.3
XP_005268559.1
15
2448
XM_005268503.3
XP_005268560.1
15
2448
XM_005268504.3
XP_005268561.1
15
2448
XM_005268505.3
XP_005268562.1
15
2448
XM_005268506.3
XP_005268563.1
15
2448
XM_005268507.3
XP_005268564.1
14
2217
XM_005268508.3
XP_005268565.1
15
2448
XM_005268509.3
XP_005268566.1
15
2448
XM_011537678.2
XP_011535980.1
15
2448
XM_017009792.1
XP_016865281.1
15
2448
XM_017009793.1
XP_016865282.1
15
2448
XM_017009794.1
XP_016865283.1
15
2448
XM_017009795.1
XP_016865284.1
14
2217
GenBank mRNA
AB209317.1
-
7
1203
AK096467.1
-
11
2017
AK225284.1
-
14
2205
AK296918.1
-
8
1227
AK299167.1
-
8
1468
AK303611.1
-
15
2436
AK307586.1
-
15
2436
AY460334.1
-
15
2403
BC008331.1
-
6
1077
BC011764.2
-
15
2432
3616898
BC014559.2
-
15
2432
4025673
BC016144.1
-
15
2431
BC027252.1
-
14
2128
BC033093.1
-
15
2432
4577077
BX537490.1
-
8
3657
U40847.1
-
14
2265
U76366.1
-
14
2247
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
transport
transporter activity
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0