Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002294.2 | NP_001002294.1 | 5 | 654 | ||
NM_001319173.1 | NP_001306102.1 | 6 | 764 | |||
NM_001319174.1 | NP_001306103.1 | 4 | 465 | |||
NM_006894.5 | NP_008825.4 | 5 | 637 | |||
XM_011509345.2 | XP_011507647.1 | 6 | 785 | |||
GenBank mRNA | AK223166.1 | - | 5 | 646 | ||
AK296105.1 | - | 4 | 439 | |||
AK313197.1 | - | 5 | 627 | |||
BC032016.1 | - | 5 | 589 | 5175615 | ||
KU178077.1 | - | 4 | 543 | |||
M83772.1 | - | 5 | 679 | |||
Z47552.1 | - | 5 | 636 |