Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128636.2 | NP_001122108.1 | 2 | 667 | ||
XM_006715725.3 | XP_006715788.1 | 3 | 834 | |||
XM_006715726.2 | XP_006715789.1 | 3 | 781 | |||
XM_006715727.3 | XP_006715790.1 | 3 | 1080 | |||
XM_011515397.2 | XP_011513699.1 | 3 | 872 | |||
XM_011515398.2 | XP_011513700.1 | 4 | 1017 | |||
XM_011515401.2 | XP_011513703.1 | 2 | 4119 | |||
XM_017012203.1 | XP_016867692.1 | 3 | 2375 | |||
GenBank mRNA | DR001365.1 | - | 1 | 416 |