Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001814.5 | NP_001805.3 | 7 | 1496 | ||
GenBank mRNA | AF234263.1 | - | 7 | 1407 | ||
AF234264.1 | - | 7 | 1407 | |||
AF254757.1 | - | 7 | 1407 | |||
AF525032.1 | - | 7 | 1407 | |||
AF525033.1 | - | 7 | 1407 | |||
AK223038.1 | - | 7 | 1445 | |||
AK292117.1 | - | 7 | 1414 | |||
AK296190.1 | - | 8 | 1394 | |||
X87212.1 | - | 7 | 1414 |