Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000194.2 | NP_000185.1 | 7 | 681 | ||
GenBank mRNA | AK313435.1 | - | 7 | 661 | ||
BC000578.2 | - | 7 | 553 | 3163726 | ||
BT019350.1 | - | 7 | 514 | |||
CR407645.1 | - | 7 | 514 | |||
L29382.1 | - | 7 | 514 | |||
L29383.1 | - | 7 | 514 | |||
M31642.1 | - | 7 | 599 |