Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006265.2 | NP_006256.1 | 2 | 365 | ||
GenBank mRNA | AK097915.1 | - | 2 | 173 | ||
AK289505.1 | - | 2 | 142 | |||
BC050381.1 | - | 2 | 205 | 6044010 | ||
D38551.1 | - | 2 | 261 | |||
DA328186.1 | - | 2 | 365 | |||
X98294.1 | - | 1 | 77 |