Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010909.3 | NP_001010909.2 | 3 | 1886 | ||
NM_001322370.1 | NP_001309299.1 | 1952 | ||||
NM_001322371.1 | NP_001309300.1 | 2059 | ||||
NR_130720.1 | 3 | 2028 | ||||
GenBank mRNA | AB242595.1 | - | 3 | 1796 | ||
AK056612.1 | - | 2 | 381 | |||
AK298642.1 | - | 3 | 1874 | |||
AK301645.1 | - | 2 | 1594 | |||
AK316363.1 | - | 3 | 1876 | |||
AY358415.1 | - | 1963 | ||||
BC105735.1 | - | 3 | 1821 | 40026480 | ||
BC105736.1 | - | 3 | 1963 | |||
BC105737.1 | - | 3 | 1821 | 40026476 | ||
BC107478.1 | - | 4 | 1371 | 40026469 | ||
HM596784.1 | - | 3 | 1815 |