Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042522.2 | NP_001035987.1 | 1, 2 | 286 | ||
XM_006723219.3 | XP_006723282.1 | 2, 3 | 277 | |||
GenBank mRNA | BC151136.1 | - | 1, 2 | 286 | 9057036 | |
DQ323927.1 | - | 1, 2 | 187 | |||
DQ323928.1 | - | 1, 2 | 187 |