Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001220484.1 | NP_001207413.1 | 12, 13 | 2632 | ||
NM_203309.2 | NP_976054.2 | 11, 12 | 2553 | |||
XM_006720143.2 | XP_006720206.1 | 11, 12 | 2590 | |||
XM_011536760.2 | XP_011535062.1 | 11, 12 | 2776 | |||
XM_017021289.1 | XP_016876778.1 | 11, 12 | 3114 | |||
GenBank mRNA | AK302564.1 | - | 12, 13 | 2606 | ||
BC047590.1 | - | 11, 12 | 2553 | 4837558 | ||
GU727638.1 | - | 10, 11 | 2143 |