Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281520.1 | NP_001268449.1 | 9 | 1362 | ||
NM_001281521.1 | NP_001268450.1 | 9 | 1308 | |||
NM_001281522.1 | NP_001268451.1 | 9 | 1308 | |||
NM_032485.5 | NP_115874.3 | 9 | 1308 | |||
NM_182802.2 | NP_877954.1 | 9 | 1362 | |||
XM_011529387.2 | XP_011527689.1 | 9 | 1271 | |||
XM_017028105.1 | XP_016883594.1 | 9 | 1337 | |||
XM_017028106.1 | XP_016883595.1 | 9 | 1274 | |||
XM_017028107.1 | XP_016883596.1 | 2 | 1547 | |||
GenBank mRNA | AJ439063.1 | - | 9 | 1308 | ||
AK027644.1 | - | 3 | 347 | |||
AK314654.1 | - | 9 | 990 | |||
AY158211.1 | - | 9 | 1061 | |||
BC005170.1 | - | 9 | 1061 | |||
BC008830.2 | - | 6 | 618 | |||
BC080656.1 | - | 9 | 1013 | 4841271 | ||
BC101054.1 | - | 9 | 1109 | 40008094 | ||
BC101055.1 | - | 9 | 1109 | 40008095 | ||
BC101056.1 | - | 9 | 1109 | 40008096 | ||
BC101057.1 | - | 9 | 1163 | 40008097 | ||
KU178803.1 | - | 3 | 220 | |||
KU178804.1 | - | 3 | 220 | |||
KU178805.1 | - | 3 | 220 |