Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276254.2 | NP_001263183.2 | 5 | 1450 | ||
NR_074079.1 | 5 | 1451 | ||||
GenBank mRNA | JN806225.1 | - | 4 | 1306 | ||
JN806226.1 | - | 3 | 1234 | |||
JN806227.1 | - | 5 | 1451 | |||
JN806228.1 | - | 4 | 1307 | |||
JN806229.1 | - | 3 | 1235 | |||
JN806231.1 | - | 4 | 946 | |||
JN806232.1 | - | 4 | 945 | |||
JN806234.1 | - | 5 | 1450 |