Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304467.1 | NP_001291396.1 | 5, 6 | 729 | ||
NM_212554.3 | NP_997719.2 | 5, 6 | 725 | |||
GenBank mRNA | AK022354.1 | - | 1, 2 | 176 | ||
AL832292.1 | - | 5, 6 | 626 | |||
BC026167.1 | - | 5, 6 | 660 | 4823003 | ||
BI551093.1 | - | 5, 6 | 630 |