Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282902.2 | NP_001269831.1 | 2 | 267 | ||
NM_001282903.2 | NP_001269832.1 | 3 | 756 | |||
NM_014112.4 | NP_054831.2 | 3 | 770 | |||
XM_005251049.2 | XP_005251106.1 | 2 | 585 | |||
XM_006716625.1 | XP_006716688.1 | 3 | 707 | |||
XM_011517264.1 | XP_011515566.1 | 4 | 840 | |||
XM_011517266.2 | XP_011515568.1 | 3 | 575 | |||
XM_011517267.1 | XP_011515569.1 | 3 | 693 | |||
XM_011517268.1 | XP_011515570.1 | 3 | 682 | |||
XM_017013801.1 | XP_016869290.1 | 2 | 4369 | |||
GenBank mRNA | AF183810.1 | - | 3 | 764 | ||
AF264784.1 | - | 2 | 311 | |||
AK304046.1 | - | 2 | 262 | |||
AK309357.1 | - | 2 | 481 | |||
BC125020.1 | - | 3 | 600 | 40073132 | ||
DC339831.1 | - | 2 | 267 |