Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000587.2 | NP_000578.2 | 6 | 663 | ||
GenBank mRNA | AK290349.1 | - | 6 | 673 | ||
AK312737.1 | - | 6 | 597 | |||
BC025402.1 | - | 6 | 742 | |||
BC063851.1 | - | 6 | 644 | 6184213 | ||
J03507.1 | - | 5 | 549 |