Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242581.1 | NP_001229510.1 | 8 | 1046 | ||
NM_018993.3 | NP_061866.1 | 7 | 786 | |||
XM_005260731.2 | XP_005260788.1 | 9 | 1242 | |||
XM_005260733.2 | XP_005260790.1 | 5 | 594 | |||
XM_006723574.3 | XP_006723637.1 | 10 | 1224 | |||
XM_006723575.3 | XP_006723638.1 | 9 | 1010 | |||
XM_006723577.2 | XP_006723640.1 | 8 | 859 | |||
XM_011529255.2 | XP_011527557.2 | 8 | 1319 | |||
XM_011529257.1 | XP_011527559.1 | 8 | 915 | |||
XM_011529258.2 | XP_011527560.1 | 8 | 927 | |||
XM_011529259.2 | XP_011527561.1 | 7 | 839 | |||
XM_017027887.1 | XP_016883376.1 | 11 | 1565 | |||
XM_017027888.1 | XP_016883377.1 | 12 | 1637 | |||
XM_017027889.1 | XP_016883378.1 | 7 | 1166 | |||
XM_017027890.1 | XP_016883379.1 | 9 | 997 | |||
XM_017027891.1 | XP_016883380.1 | 9 | 1035 | |||
XM_017027892.1 | XP_016883381.1 | 7 | 850 | |||
XM_017027893.1 | XP_016883382.1 | 8 | 1344 | |||
GenBank mRNA | AB060339.1 | - | 7 | 750 | ||
AK094884.1 | - | 8 | 1047 | |||
AL136924.1 | - | 6 | 667 |