Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172250.2 | NP_758454.1 | 2 | 337 | ||
XM_011531684.2 | XP_011529986.1 | 2 | 415 | |||
XM_011531685.2 | XP_011529987.1 | 2 | 458 | |||
XM_011531686.2 | XP_011529988.1 | 1 | 317 | |||
GenBank mRNA | AK126662.1 | - | 2 | 337 | ||
BC101178.2 | - | 1 | 299 | 40017262 | ||
BC101179.2 | - | 1 | 299 | 40017263 | ||
BC101180.2 | - | 1 | 299 | |||
BC101181.2 | - | 1 | 299 | 40017265 |