Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032898.4 | NP_116287.2 | 3 | 837 | ||
XM_005269370.4 | XP_005269427.1 | 3 | 812 | |||
XM_011513246.2 | XP_011511548.1 | 3 | 573 | |||
GenBank mRNA | AK058155.1 | - | 3 | 740 | ||
AK314070.1 | - | 3 | 806 | |||
AY099509.1 | - | 3 | 767 | |||
BC007827.2 | - | 3 | 766 | 3533603 |