Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190879.2 | NP_001177808.1 | 12 | 1223 | ||
NM_001190881.2 | NP_001177810.1 | 8 | 841 | |||
NM_001190882.2 | NP_001177811.1 | 10 | 985 | |||
NM_002552.4 | NP_002543.2 | 11 | 1107 | |||
NM_181741.3 | NP_859525.1 | 11 | 1216 | |||
NM_181742.3 | NP_859526.1 | 11 | 1111 | |||
XM_011511255.2 | XP_011509557.1 | 12 | 1261 | |||
XM_017004226.1 | XP_016859715.1 | 13 | 1321 | |||
XM_017004227.1 | XP_016859716.1 | 10 | 945 | |||
GenBank mRNA | AF022108.1 | - | 11 | 1063 | ||
AF047598.1 | - | 11 | 1062 | |||
AF132596.1 | - | 11 | 1049 | |||
AK225092.1 | - | 11 | 1024 | |||
AK291989.1 | - | 11 | 1081 | |||
AK294878.1 | - | 11 | 1059 | |||
AK295721.1 | - | 8 | 810 | |||
AK298862.1 | - | 10 | 985 | |||
AK299749.1 | - | 9 | 846 | |||
BC005388.1 | - | 11 | 1028 | |||
BC014847.1 | - | 11 | 1050 | 3446657 | ||
BC016177.1 | - | 7 | 699 |