Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142301.1 | NP_001135773.1 | 16 | 1568 | ||
NM_153704.5 | NP_714915.3 | 15 | 1637 | |||
NR_024522.1 | 15 | 1637 | ||||
XM_006716686.3 | XP_006716749.1 | 14 | 1475 | |||
XM_011517363.2 | XP_011515665.1 | 6 | 727 | |||
GenBank mRNA | AK026742.1 | - | 10 | 943 | ||
AK092244.1 | - | 16 | 1525 | |||
AK094935.1 | - | 15 | 1561 | |||
AK308735.1 | - | 6 | 704 | |||
BC032835.2 | - | 15 | 1555 | 4825770 | ||
BC054338.1 | - | 15 | 1564 | |||
BX648768.1 | - | 15 | 1581 | |||
KU178827.1 | - | 15 | 1536 | |||
KU178828.1 | - | 14 | 1382 |