Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142301.1 | NP_001135773.1 | 26 | 2627 | ||
NM_153704.5 | NP_714915.3 | 25 | 2696 | |||
NR_024522.1 | 25 | 2696 | ||||
XM_006716686.3 | XP_006716749.1 | 24 | 2534 | |||
XM_011517363.2 | XP_011515665.1 | 16 | 1786 | |||
GenBank mRNA | AK026742.1 | - | 20 | 2002 | ||
AK092244.1 | - | 26 | 2584 | |||
AK094935.1 | - | 25 | 2620 | |||
AK297676.1 | - | 10 | 1349 | |||
AK308735.1 | - | 16 | 1763 | |||
BC032835.2 | - | 25 | 2614 | 4825770 | ||
BC054338.1 | - | 25 | 2623 | |||
BX648768.1 | - | 25 | 2640 | |||
KU178827.1 | - | 25 | 2595 | |||
KU178828.1 | - | 24 | 2441 | |||
KU178829.1 | - | 6 | 499 |