Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014028.3 | NP_054747.2 | 3 | 690 | ||
GenBank mRNA | AF077205.1 | - | 1 | 42 | ||
AF533891.1 | - | 3 | 674 | |||
AK075012.1 | - | 3 | 616 | |||
AK129854.1 | - | 3 | 684 | |||
AK314393.1 | - | 3 | 366 | |||
AY358795.1 | - | 3 | 681 | |||
BC068581.1 | - | 3 | 672 | 5298450 |