Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014028.3 | NP_054747.2 | 4, 5 | 875 | ||
GenBank mRNA | AF077205.1 | - | 2, 3 | 228 | ||
AF533891.1 | - | 4, 5 | 859 | |||
AK075012.1 | - | 4, 5 | 801 | |||
AK129854.1 | - | 4, 5 | 869 | |||
AK314393.1 | - | 4, 5 | 551 | |||
AY358795.1 | - | 4, 5 | 866 | |||
BC068581.1 | - | 4, 5 | 857 | 5298450 |