Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAMLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177465.2 | NP_001170936.1 | 4 | 2218 | ||
NM_001177466.2 | NP_001170937.1 | 5 | 2332 | |||
NM_005491.4 | NP_005482.2 | 6 | 2407 | |||
XM_006724802.3 | XP_006724865.1 | 8 | 2755 | |||
XM_006724803.3 | XP_006724866.1 | 7 | 2680 | |||
XM_011531092.2 | XP_011529394.1 | 6 | 2388 | |||
XM_011531093.2 | XP_011529395.1 | 7 | 2518 | |||
XM_011531095.2 | XP_011529397.1 | 7 | 2632 | |||
XM_017029188.1 | XP_016884677.1 | 8 | 2627 | |||
XM_017029189.1 | XP_016884678.1 | 7 | 2505 | |||
GenBank mRNA | AK094563.1 | - | 1 | 697 | ||
AK125184.1 | - | 1 | 1727 | |||
AK294478.1 | - | 4 | 2077 | |||
AK297504.1 | - | 3 | 788 | |||
AK302612.1 | - | 8 | 2179 | |||
BC136324.1 | - | 5 | 2317 | 9020311 | ||
BX537560.1 | - | 1 | 4078 | |||
DA527328.1 | - | 4 | 486 | |||
U46023.1 | - | 6 | 2408 |