Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOXP1 SIRNA Assays ›
Gene Symbol:
FOXP1
Gene Name:
forkhead box P1
Gene Aliases:
12CC4, HSPC215, MFH, QRF1, hFKH1B
Chromosome Location:
Chr.3: 70954714 - 71583989 on Build GRCh38
UniGene ID:
Hs.59368
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s532468
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOXP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001244808.1
NP_001231737.1
12
1592
NM_001244810.1
NP_001231739.1
12
1489
NM_001244812.1
NP_001231741.1
7
889
NM_001244813.1
NP_001231742.1
6
781
NM_001244814.1
NP_001231743.1
8
1286
NM_001244815.1
NP_001231744.1
7
1227
NM_001244816.1
NP_001231745.1
11
1394
NM_032682.5
NP_116071.2
12
1489
XM_005264735.3
XP_005264792.1
7
1175
XM_005264736.3
XP_005264793.1
7
1227
XM_005264737.4
XP_005264794.1
12
1495
XM_005264742.3
XP_005264799.1
6
781
XM_006713102.2
XP_006713165.1
11
1450
XM_006713103.2
XP_006713166.1
9
1187
XM_006713104.2
XP_006713167.1
10
1420
XM_011533584.2
XP_011531886.1
12
1560
XM_011533585.2
XP_011531887.1
11
1412
XM_011533588.2
XP_011531890.1
7
1010
XM_017006165.1
XP_016861654.1
11
1331
XM_017006166.1
XP_016861655.1
11
1427
XM_017006167.1
XP_016861656.1
7
1224
GenBank mRNA
AB052767.1
-
2
226
AF146696.1
-
10
1226
AF250920.1
-
11
1394
AF275309.1
-
3
337
AK027264.1
-
7
859
AK074321.1
-
8
1285
AK074569.1
-
6
781
AK092383.1
-
7
889
AK122710.1
-
12
1592
AK297649.1
-
7
692
BC131720.1
-
9
1111
BX647682.1
-
6
679
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of B cell apoptotic process
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
positive regulation of endothelial cell migration
osteoclast differentiation
response to lipopolysaccharide
regulation of tumor necrosis factor production
chemokine (C-C motif) ligand 2 secretion
osteoclast development
macrophage activation
monocyte activation
endothelial cell activation
regulation of monocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of smooth muscle cell proliferation
regulation of interleukin-1 beta secretion
regulation of inflammatory response
negative regulation of androgen receptor signaling pathway
T follicular helper cell differentiation
interleukin-21 secretion
regulation of defense response to bacterium
regulation of macrophage colony-stimulating factor production
regulation of endothelial tube morphogenesis
regulation of interleukin-12 secretion
RNA polymerase II transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
protein self-association
metal ion binding
androgen receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0