Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005252066.3 | XP_005252123.1 | 2, 3 | 349 | ||
XM_005252067.4 | XP_005252124.1 | 2, 3 | 351 | |||
XM_005252069.4 | XP_005252126.1 | 2, 3 | 352 | |||
XM_006717150.3 | XP_006717213.1 | 2, 3 | 352 | |||
XM_011518800.2 | XP_011517102.1 | 2, 3 | 353 | |||
XM_011518803.2 | XP_011517105.1 | 2, 3 | 357 |