Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000254.2 | NP_000245.2 | 18 | 2359 | ||
NM_001291939.1 | NP_001278868.1 | 18 | 2359 | |||
NM_001291940.1 | NP_001278869.1 | 17 | 2246 | |||
XM_005273141.4 | XP_005273198.1 | 18 | 2328 | |||
XM_006711770.2 | XP_006711833.1 | 12 | 1307 | |||
XM_011544194.2 | XP_011542496.1 | 17 | 3121 | |||
XM_017001329.1 | XP_016856818.1 | 17 | 3121 | |||
XM_017001330.1 | XP_016856819.1 | 17 | 3121 | |||
GenBank mRNA | AK307390.1 | - | 18 | 2348 | ||
BC130616.1 | - | 18 | 1958 | 40146647 | ||
BC136440.1 | - | 18 | 1958 | 9020429 | ||
BC144095.1 | - | 18 | 1958 | 9052615 | ||
BC144097.1 | - | 18 | 1958 | 9052617 | ||
BC144098.1 | - | 17 | 1845 | |||
BX641132.1 | - | 5 | 598 | |||
U71285.1 | - | 18 | 1999 | |||
U73338.1 | - | 18 | 2330 | |||
U75743.1 | - | 18 | 2222 |