Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135031.1 | NP_001128503.1 | 4 | 473 | ||
NM_004188.6 | NP_004179.3 | 8 | 1147 | |||
XM_006717297.3 | XP_006717360.1 | 4 | 638 | |||
XM_011519068.2 | XP_011517370.1 | 6 | 1038 | |||
XM_011519069.2 | XP_011517371.1 | 5 | 808 | |||
XM_011519070.2 | XP_011517372.1 | 6 | 1038 | |||
XM_017015175.1 | XP_016870664.1 | 5 | 768 | |||
XM_017015176.1 | XP_016870665.1 | 6 | 1038 | |||
GenBank mRNA | AF081946.1 | - | 3 | 322 | ||
AY428733.1 | - | 4 | 473 | |||
BC043371.1 | - | 8 | 1147 | 5170920 | ||
BC048254.1 | - | 3 | 353 | |||
CR536546.1 | - | 3 | 322 |