Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000405.4 | NP_000396.2 | 4 | 886 | ||
NM_001167607.1 | NP_001161079.1 | 4 | 676 | |||
GenBank mRNA | AK298867.1 | - | 4 | 543 | ||
BC009273.2 | - | 4 | 750 | 4053681 | ||
L01439.1 | - | 4 | 811 | |||
M76477.1 | - | 4 | 811 | |||
X16087.1 | - | 4 | 679 | |||
X61095.1 | - | 4 | 742 | |||
X62078.1 | - | 4 | 779 |