Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002028.3 | NP_002019.1 | 12 | 1407 | ||
GenBank mRNA | AK093298.1 | - | 10 | 1165 | ||
AK225917.1 | - | 12 | 1274 | |||
AK295972.1 | - | 9 | 971 | |||
AK315714.1 | - | 12 | 1474 | |||
BC020232.1 | - | 12 | 1257 | 4652202 | ||
BX248269.1 | - | 12 | 1253 | |||
L00635.1 | - | 12 | 1261 | |||
L10414.1 | - | 11 | 1092 |
Gene Symbol: CHURC1-FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202558.1 | NP_001189487.1 | 13 | 1408 | ||
NM_001202559.1 | NP_001189488.1 | 14 | 1479 | |||
GenBank mRNA | AK296850.1 | - | 14 | 1347 | ||
AK303739.1 | - | 13 | 1284 |