Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161498.1 | NP_001154970.1 | 1, 2 | 366 | ||
GenBank mRNA | AK126770.1 | - | 1, 2 | 428 | ||
BC146872.1 | - | 1, 2 | 271 | 9056800 |