Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006946.2 | NP_008877.1 | 8 | 877 | ||
XM_005274192.4 | XP_005274249.1 | 9 | 1539 | |||
XM_005274193.3 | XP_005274250.1 | 8 | 1313 | |||
XM_006718669.3 | XP_006718732.1 | 7 | 3226 | |||
XM_006718671.3 | XP_006718734.1 | 9 | 939 | |||
XM_011545216.2 | XP_011543518.1 | 7 | 3236 | |||
XM_011545217.2 | XP_011543519.1 | 7 | 3236 | |||
XM_017018174.1 | XP_016873663.1 | 9 | 1631 | |||
XM_017018175.1 | XP_016873664.1 | 9 | 1260 | |||
XM_017018176.1 | XP_016873665.1 | 9 | 980 | |||
XM_017018177.1 | XP_016873666.1 | 8 | 1169 | |||
XM_017018178.1 | XP_016873667.1 | 8 | 852 | |||
GenBank mRNA | AB008567.1 | - | 8 | 877 | ||
BC139788.1 | - | 8 | 848 |