Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005554.3 | NP_005545.1 | 9 | 2437 | ||
GenBank mRNA | BC008807.2 | - | 9 | 2276 | 3947610 | |
BC014152.2 | - | 9 | 2280 | 3639270 | ||
BG059253.1 | - | 1 | 120 |