Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 11 | 1853 | ||
NM_012470.3 | NP_036602.1 | 11 | 1853 | |||
NR_034053.2 | 12 | 2014 | ||||
XM_011515989.2 | XP_011514291.1 | 11 | 1455 | |||
GenBank mRNA | AF145029.1 | - | 11 | 1665 | ||
AJ133769.1 | - | 11 | 1450 | |||
AK001263.1 | - | 4 | 407 | |||
AK022910.1 | - | 2 | 187 | |||
AK225999.1 | - | 11 | 1824 | |||
AK312580.1 | - | 11 | 1580 | |||
BC068530.1 | - | 11 | 1581 |