Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013334.3 | NP_037466.2 | 6, 7 | 895 | ||
NM_021971.2 | NP_068806.1 | 6, 7 | 895 | |||
GenBank mRNA | AF135421.1 | - | 6, 7 | 787 | ||
AK024319.1 | - | 6, 7 | 872 | |||
AK291700.1 | - | 6, 7 | 870 | |||
BC001141.1 | - | 6, 7 | 834 | 3503869 | ||
BC008033.1 | - | 6, 7 | 834 | 3355099 |