Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103738.1 | 7 | 2974 | |||
GenBank mRNA | AK056229.1 | - | 7 | 1994 | ||
AK094963.1 | - | 7 | 2975 |