Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207459.3 | NP_997342.1 | 2 | 1696 | ||
XM_017024638.1 | XP_016880127.1 | 2 | 1755 | |||
XM_017024639.1 | XP_016880128.1 | 2 | 1783 | |||
GenBank mRNA | AK093086.1 | - | 2 | 1699 | ||
BC016939.2 | - | 1 | 1657 | 4419895 | ||
BC036191.1 | - | 2 | 1696 | 5297531 | ||
BC050391.1 | - | 2 | 1673 | 5745215 | ||
BC057820.1 | - | 2 | 1697 | 4837469 |