Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIF1A SIRNA Assays ›
Gene Symbol:
KIF1A
Gene Name:
kinesin family member 1A
Gene Aliases:
ATSV, C2orf20, HSN2C, MRD9, SPG30, UNC104
Chromosome Location:
Chr.2: 240713764 - 240821025 on Build GRCh38
UniGene ID:
Hs.516802
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533731
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIF1A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001244008.1
NP_001230937.1
3
393
NM_001320705.1
NP_001307634.1
3
393
NM_004321.6
NP_004312.2
2
206
XM_005247022.1
XP_005247079.1
3
304
XM_005247024.1
XP_005247081.1
3
304
XM_005247027.1
XP_005247084.1
3
304
XM_005247028.2
XP_005247085.1
3
307
XM_006712605.1
XP_006712668.1
3
304
XM_011511364.1
XP_011509666.1
3
316
XM_011511365.1
XP_011509667.1
3
304
XM_017004383.1
XP_016859872.1
3
304
XM_017004384.1
XP_016859873.1
3
316
XM_017004385.1
XP_016859874.1
3
304
XM_017004386.1
XP_016859875.1
3
304
XM_017004387.1
XP_016859876.1
3
304
XM_017004388.1
XP_016859877.1
3
304
XM_017004389.1
XP_016859878.1
3
304
XM_017004390.1
XP_016859879.1
3
304
XM_017004391.1
XP_016859880.1
2
158
GenBank mRNA
AB210029.1
-
3
393
AB290172.1
-
3
393
AB621817.1
-
3
320
AI815863.1
-
3
292
AI878864.1
-
3
292
BC064906.1
-
3
296
BC111780.1
-
2
174
40037563
BC111799.1
-
2
174
40037561
KJ534874.1
-
2
146
KJ534875.1
-
2
146
KJ534994.1
-
2
146
KJ535053.1
-
2
146
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
anterograde axonal transport
motor activity
microtubule motor activity
ATP binding
microtubule binding
ATPase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-sjrww:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0