Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000202.7 | NP_000193.1 | 9 | 1703 | ||
NM_001166550.3 | NP_001160022.1 | 9 | 1659 | |||
GenBank mRNA | AK294541.1 | - | 9 | 1614 | ||
AK295815.1 | - | 2 | 346 | |||
BX647357.1 | - | 9 | 1557 | |||
M58342.1 | - | 9 | 1610 |