Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020774.3 | NP_065825.1 | 8 | 1380 | ||
XM_017025873.1 | XP_016881362.1 | 5 | 1072 | |||
XM_017025874.1 | XP_016881363.1 | 8 | 1916 | |||
XM_017025875.1 | XP_016881364.1 | 8 | 1916 | |||
GenBank mRNA | AK091266.1 | - | 7 | 810 | ||
AY147849.1 | - | 8 | 1398 | |||
AY149908.1 | - | 8 | 1125 | |||
BC110581.1 | - | 8 | 1125 | 40008394 | ||
BC110582.1 | - | 8 | 1125 | 40008395 | ||
CR749635.1 | - | 5 | 545 |