Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305581.1 | NP_001292510.1 | 6 | 669 | ||
NM_032024.4 | NP_114413.1 | 5 | 721 | |||
NR_131178.1 | 7 | 944 | ||||
XM_011540256.2 | XP_011538558.1 | 6 | 663 | |||
XM_017016759.1 | XP_016872248.1 | 6 | 663 | |||
XM_017016760.1 | XP_016872249.1 | 6 | 663 | |||
XM_017016761.1 | XP_016872250.1 | 6 | 663 | |||
XM_017016762.1 | XP_016872251.1 | 2 | 499 | |||
GenBank mRNA | AF267860.1 | - | 5 | 722 | ||
AK125328.1 | - | 2 | 952 | |||
BX105111.1 | - | 5 | 563 | |||
CR933639.1 | - | 7 | 944 |