Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191060.1 | NP_001177989.1 | 9 | 1346 | ||
NM_001191061.1 | NP_001177990.1 | 9 | 1015 | |||
NM_024698.5 | NP_078974.1 | 9 | 1183 | |||
XM_011520369.1 | XP_011518671.1 | 9 | 1057 | |||
XM_011520370.1 | XP_011518672.1 | 9 | 1125 | |||
XM_011520371.1 | XP_011518673.1 | 9 | 1010 | |||
GenBank mRNA | AJ428202.1 | - | 8 | 903 | ||
AK023106.1 | - | 8 | 903 | |||
AK290481.1 | - | 9 | 1346 | |||
AL832423.1 | - | 4 | 435 | |||
BC019033.2 | - | 9 | 981 | 4577143 | ||
BC023545.2 | - | 9 | 1141 | 4110556 | ||
BC024212.2 | - | 5 | 524 |