Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168370.1 | NP_001161842.1 | 19 | 3965 | ||
NM_014780.4 | NP_055595.2 | 19 | 3958 | |||
XM_005249503.2 | XP_005249560.1 | 19 | 3821 | |||
XM_006715285.1 | XP_006715348.1 | 19 | 4059 | |||
XM_011515019.2 | XP_011513321.1 | 19 | 3916 | |||
XM_011515020.2 | XP_011513322.1 | 19 | 3819 | |||
XM_011515021.1 | XP_011513323.1 | 14 | 2000 | |||
XM_017011533.1 | XP_016867022.1 | 19 | 3944 | |||
XM_017011534.1 | XP_016867023.1 | 19 | 3943 | |||
XM_017011535.1 | XP_016867024.1 | 19 | 3847 | |||
XM_017011536.1 | XP_016867025.1 | 19 | 4086 | |||
XM_017011537.1 | XP_016867026.1 | 19 | 4059 | |||
XM_017011538.1 | XP_016867027.1 | 19 | 3990 | |||
XM_017011539.1 | XP_016867028.1 | 19 | 3963 | |||
GenBank mRNA | AK292806.1 | - | 19 | 3895 | ||
AK302668.1 | - | 19 | 3902 | |||
AK307936.1 | - | 9 | 1386 | |||
AK309902.1 | - | 9 | 1563 | |||
BC033647.1 | - | 19 | 3940 | 5580027 | ||
D38548.1 | - | 19 | 3712 |