Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001927.3 | NP_001918.3 | 4 | 943 | ||
GenBank mRNA | AF055081.1 | - | 4 | 857 | ||
AF055082.1 | - | 4 | 857 | |||
AF055083.1 | - | 4 | 857 | |||
AF137053.1 | - | 4 | 857 | |||
AF167579.1 | - | 4 | 857 | |||
AF486807.1 | - | 4 | 857 | |||
AF487828.1 | - | 4 | 857 | |||
AF521879.1 | - | 4 | 857 | |||
AF527578.1 | - | 4 | 857 | |||
AK022087.1 | - | 1133 | ||||
AK097038.1 | - | 5 | 883 | |||
AK098332.1 | - | 5 | 737 | |||
AK123787.1 | - | 5 | 682 | |||
AL541778.3 | - | 2 | 219 | |||
AY083345.1 | - | 4 | 857 | |||
AY125465.1 | - | 4 | 857 | |||
BC010072.2 | - | 4 | 927 | |||
BC032116.2 | - | 4 | 955 | 4905678 | ||
DQ104335.1 | - | 4 | 857 | |||
DQ104336.1 | - | 4 | 857 | |||
DQ104337.1 | - | 4 | 857 | |||
EF617312.1 | - | 4 | 937 | |||
JX114780.1 | - | 4 | 857 | |||
U59167.1 | - | 4 | 934 |