Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DDX11 SIRNA Assays ›
Gene Symbol:
DDX11
Gene Name:
DEAD/H-box helicase 11
Gene Aliases:
CHL1, CHLR1, KRG2, WABS
Chromosome Location:
Chr.12: 31073574 - 31104799 on Build GRCh38
UniGene ID:
Hs.443960
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s534392
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DDX11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001257144.1
NP_001244073.1
27
3775
NM_001257145.1
NP_001244074.1
27
3707
NM_004399.2
NP_004390.3
26
3752
NM_030653.3
NP_085911.2
27
3902
NM_152438.1
NP_689651.1
27
3897
XM_005253331.3
XP_005253388.1
27
4067
XM_005253333.3
XP_005253390.1
27
4072
XM_006719050.3
XP_006719113.1
26
4147
XM_011520592.2
XP_011518894.1
26
4143
XM_011520595.2
XP_011518897.1
26
3922
XM_017018918.1
XP_016874407.1
27
4325
XM_017018919.1
XP_016874408.1
28
4388
XM_017018920.1
XP_016874409.1
27
4287
XM_017018921.1
XP_016874410.1
28
4250
XM_017018922.1
XP_016874411.1
27
4118
XM_017018923.1
XP_016874412.1
27
4011
XM_017018924.1
XP_016874413.1
27
4044
XM_017018925.1
XP_016874414.1
27
4116
XM_017018926.1
XP_016874415.1
27
4185
XM_017018927.1
XP_016874416.1
26
4176
XM_017018928.1
XP_016874417.1
27
4330
XM_017018929.1
XP_016874418.1
27
4292
XM_017018930.1
XP_016874419.1
28
4254
XM_017018931.1
XP_016874420.1
26
3943
XM_017018932.1
XP_016874421.1
27
3761
XM_017018933.1
XP_016874422.1
27
4412
XM_017018934.1
XP_016874423.1
25
3864
XM_017018935.1
XP_016874424.1
23
3348
XM_017018936.1
XP_016874425.1
23
4343
XM_017018937.1
XP_016874426.1
23
4348
GenBank mRNA
BC050069.1
-
27
3692
5271949
BC050522.1
-
27
3726
6471750
X99583.1
-
26
3712
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA helicase
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
sister chromatid cohesion
viral process
DNA duplex unwinding
IRE1-mediated unfolded protein response
double-stranded DNA binding
single-stranded DNA binding
RNA binding
ATP-dependent DNA helicase activity
helicase activity
protein binding
ATP binding
DNA-dependent ATPase activity
metal ion binding
4 iron, 4 sulfur cluster binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0