Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324281.1 | NP_001311210.1 | 19 | 2808 | ||
NM_032415.5 | NP_115791.3 | 18 | 2747 | |||
XM_011515586.2 | XP_011513888.1 | 19 | 2776 | |||
XM_011515587.2 | XP_011513889.1 | 18 | 2739 | |||
GenBank mRNA | AF322641.1 | - | 18 | 2649 | ||
AF352576.1 | - | 16 | 2322 | |||
AK074049.1 | - | 18 | 2667 | |||
BC111719.1 | - | 17 | 2433 | 40009168 |