Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015346.3 | NP_056161.2 | 23 | 4790 | ||
XM_011536609.2 | XP_011534911.1 | 23 | 4791 | |||
XM_017021124.1 | XP_016876613.1 | 23 | 4791 | |||
XM_017021125.1 | XP_016876614.1 | 23 | 4791 | |||
XM_017021126.1 | XP_016876615.1 | 22 | 5405 | |||
XM_017021127.1 | XP_016876616.1 | 12 | 2487 | |||
XM_017021128.1 | XP_016876617.1 | 12 | 2340 | |||
GenBank mRNA | AB002319.1 | - | 8 | 1660 | ||
AB425197.1 | - | 23 | 4778 | |||
BC144594.1 | - | 21 | 4094 | |||
BX537886.1 | - | 23 | 4784 | |||
BX538025.1 | - | 19 | 4283 | |||
BX648683.1 | - | 23 | 4788 | |||
CR749276.1 | - | 24 | 4725 | |||
CR749365.1 | - | 23 | 4789 |