Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015346.3 | NP_056161.2 | 36 | 6743 | ||
XM_017021124.1 | XP_016876613.1 | 36 | 6762 | |||
XM_017021125.1 | XP_016876614.1 | 36 | 6762 | |||
XM_017021126.1 | XP_016876615.1 | 35 | 7376 | |||
XM_017021127.1 | XP_016876616.1 | 25 | 4458 | |||
XM_017021128.1 | XP_016876617.1 | 25 | 4311 | |||
GenBank mRNA | AB002319.1 | - | 21 | 3613 | ||
AB425197.1 | - | 36 | 6731 | |||
AK024743.1 | - | 8 | 1105 | |||
BC008927.2 | - | 2 | 142 | |||
BC033235.1 | - | 3 | 438 | |||
BC144594.1 | - | 34 | 6047 | |||
BX537886.1 | - | 36 | 6737 | |||
BX538025.1 | - | 32 | 6236 | |||
CR749276.1 | - | 37 | 6678 | |||
CR749365.1 | - | 35 | 7356 |