Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146289.1 | NP_001139761.1 | 6 | 1224 | ||
NM_001243246.1 | NP_001230175.1 | 6 | 1224 | |||
NM_022356.3 | NP_071751.3 | 6 | 1224 | |||
XM_005271110.2 | XP_005271167.1 | 5 | 561 | |||
XM_011541948.1 | XP_011540250.1 | 6 | 616 | |||
XM_017002051.1 | XP_016857540.1 | 7 | 1196 | |||
XM_017002052.1 | XP_016857541.1 | 7 | 1193 | |||
GenBank mRNA | AF097431.1 | - | 7 | 1217 | ||
AF097432.1 | - | 6 | 1162 | |||
AK025841.1 | - | 2 | 80 | |||
AK027648.1 | - | 6 | 1152 | |||
AK027680.1 | - | 6 | 1126 | |||
AK027697.1 | - | 6 | 1163 | |||
AK075418.1 | - | 6 | 1141 | |||
AK297980.1 | - | 7 | 1029 | |||
AK300210.1 | - | 7 | 1115 | |||
BC015309.2 | - | 2 | 126 | |||
BC108311.1 | - | 6 | 1162 | 4521167 | ||
BC143961.1 | - | 6 | 1140 | |||
BT007039.1 | - | 2 | 73 |