Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024334.2 | NP_077310.1 | 2 | 403 | ||
XM_017007176.1 | XP_016862665.1 | 3 | 525 | |||
GenBank mRNA | AK027757.1 | - | 2 | 329 | ||
AK027877.1 | - | 2 | 267 | |||
AK075010.1 | - | 2 | 254 | |||
AK225589.1 | - | 2 | 326 | |||
AL136916.1 | - | 2 | 297 | |||
AY358625.1 | - | 2 | 166 | |||
BC003125.1 | - | 2 | 298 | 3503636 | ||
BC011719.2 | - | 2 | 298 | 4079766 | ||
DA259919.1 | - | 2 | 408 | |||
KU178769.1 | - | 1 | 149 | |||
KU178770.1 | - | 1 | 149 |